missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetonitrile, Gradient HPLC Grade, Scharlab
Click to view available options
Quantity:
2.5 L
Packaging:
Amber Glass Bottle
Unit Size:
2.5 L
Description
- Undergoes highly efficient distillation, solid phase absorption and other treatments to eliminate impurities that could interfere in HPLC analysis
- Microfiltered and packed in amber-coloured bottles (sealed with a PTFE membrane in the cap) to avoid possible contamination
Specifications
Specifications
| Melting Point | -45.7°C |
| CAS | 75-05-8 |
| Color | Colorless |
| Density | 0.786g/cm3 |
| Boiling Point | 81.6°C |
| Quantity | 2.5 L |
| Flash Point | 2°C |
| Ignition Point | 524°C |
| Packaging | Amber Glass Bottle |
| Physical Form | Liquid |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction