missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Acid Fast Bacillus Stain Kits
Shop All Epredia Products
Click to view available options
Type:
Carbol Fuchsin
Differentiating Solution
Fast Green Stain Solution
Loeffler Methylene Blue Stain Solution
Unit Size:
125mL
250mL
500mL
Description
Acid-fast bacilli are stained bright red against a pale blue or pale green background.
Green Secondary Stain Kit includes:
- Fast Green Stain Solution, 250mL
- Carbol Fuchsin Solution, 125mL
- Differentiating Solution, 500mL
Blue Secondary Stain Kit includes:
- Loeffler Methylene Blue Stain Solution
- Carbol Fuchsin Solution, 125mL
- Differentiating Solution, 500mL
Specifications
Specifications
| Type | Loeffler Methylene Blue Stain Solution |
Safety and Handling
Signal Word
- Warning
- LONG-TERM AQUATIC HAZARD Chronic 2
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H411-Toxic to aquatic life with long lasting effects.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P362-Take off contaminated clothing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- MIXTURE LIST-Contains : paraffin waxes and hydrocarbon waxes, polyisobutylene, 2,6-ditertbutyl-p-cresol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction