Learn More
Cytiva (formerly Pall Lab) Acrodisc™ Syringe Filters with Fluorodyne™ II Membrane Optimized for Scale Up
Fluorodyne II membrane is a hydrophilic polyvinylidene fluoride (PVDF) that offers higher flow rates with low protein and preservative binding, and broad chemical and temperature resistance.
Brand: Cytiva (formerly Pall Lab) 4907
Additional Details : Weight : 0.19000kg
Description
- Higher flow rates with low protein and preservative binding
- Broad chemical and temperature resistance
Specifications
PVDF | |
0.2 μm | |
Sterile |