Learn More
Cytiva (formerly Pall Lab) AcroPrep™ Advance 96-well Filter Plates for Solvent Filtration
Optimized for a variety of applications requiring larger per well volume processing.
200.00 EUR - 432.00 EUR
Specifications
Sterility Non-sterile
For Use With (Application) Solvent Filtration
View More Specs
Products 14
Product Code Brand Volume (Metric) Pore Size Material (Membrane) Price Quantity & Availability  
Product Code Brand Volume (Metric) Pore Size Material (Membrane) Price Quantity & Availability  
17134611
View Documents Promotion Details
Cytiva (formerly Pall Lab)
8047
350 μL 0.2 μm PTFE
312.00 EUR
Pack of 10
View alternative products
Save Up to 
»
Estimated Shipment: 21-08-2024
to see stock.
Add to basket
17114621
View Documents Promotion Details
Cytiva (formerly Pall Lab)
8129
1 mL 0.45 μm Supor (PES)
213.00 EUR
Pack of 5
View alternative products
Save Up to 
»
Estimated Shipment: 12-08-2024
to see stock.
Add to basket
17124621
View Documents Promotion Details
Cytiva (formerly Pall Lab)
8130
1 mL 1.2 μm Supor (PES)
213.00 EUR
Pack of 5
View alternative products
Save Up to 
»
Estimated Shipment: 12-08-2024
to see stock.
Add to basket
17144611
View Documents Promotion Details
Cytiva (formerly Pall Lab)
8048
350 μL 0.45 μm PTFE
308.00 EUR
Pack of 10
View alternative products
Save Up to 
»
Estimated Shipment: 12-08-2024
to see stock.
Add to basket
17174631
View Documents Promotion Details
Cytiva (formerly Pall Lab)
8247
2 mL 0.2 μm PTFE
234.00 EUR
Pack of 5
View alternative products
Save Up to 
»
Estimated Shipment: 16-07-2024
to see stock.
Add to basket
17164621
View Documents Promotion Details
Cytiva (formerly Pall Lab)
8147
1 mL 0.2 μm PTFE
403.00 EUR
Pack of 5
View alternative products
Save Up to 
»
Estimated Shipment: 09-09-2024
to see stock.
Add to basket
17174621
View Documents Promotion Details
Cytiva (formerly Pall Lab)
8148
1 mL 0.45 μm PTFE
432.00 EUR
Pack of 5
View alternative products
Save Up to 
»
Estimated Shipment: 16-07-2024
to see stock.
Add to basket
17104641
View Documents Promotion Details
Cytiva (formerly Pall Lab)
8582
350 μL 0.2 μm wwPTFE
266.00 EUR
Pack of 10
View alternative products
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 12-08-2024
to see stock.
Add to basket
17144641
missing translation for 'viewDocuments' Promotion Details
Cytiva (formerly Pall Lab)
8682
1 mL 0.2 μm wwPTFE
200.00 EUR
Pack of 5
View alternative products
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 10-06-2024
to see stock.
Add to basket
17154641
missing translation for 'viewDocuments' Promotion Details
Cytiva (formerly Pall Lab)
8684
1 mL 0.45 μm wwPTFE
204.00 EUR
Pack of 5
View alternative products
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 24-06-2024
to see stock.
Add to basket
17114641
missing translation for 'viewDocuments' Promotion Details
Cytiva (formerly Pall Lab)
8584
350 μL 0.45 μm wwPTFE
266.00 EUR
Pack of 10
View alternative products
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 12-08-2024
to see stock.
Add to basket
17184631
missing translation for 'viewDocuments' Promotion Details
Cytiva (formerly Pall Lab)
8248
2 mL 0.45 μm PTFE
234.00 EUR
Pack of 5
View alternative products
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 16-07-2024
to see stock.
Add to basket
17184641
missing translation for 'viewDocuments' Promotion Details
Cytiva (formerly Pall Lab)
8782
2 mL 0.65/0.2 μm wwPTFE
230.00 EUR
Pack of 5
View alternative products
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 09-09-2024
to see stock.
Add to basket
17194641
missing translation for 'viewDocuments' Promotion Details
Cytiva (formerly Pall Lab)
8784
2 mL 0.45 μm wwPTFE
204.00 EUR
Pack of 5
View alternative products
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 24-06-2024
Description
  • Plates are constructed of chemically resistant, biologically-inert polypropylene
  • This product is not approved for use in medical, clinical, surgical or other patient applications
Specifications
Specifications
Non-sterile
Solvent Filtration