missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Acrosomal Vesicle Protein Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA558512
This item is not returnable.
View return policy
Description
Immunogen sequence: SGEHASGEQA SGAPISSTST GTILNCYTCA YMNDQGKCLR GEGTCITQNS QQCMLKKIFE GGKLQFMVQG CENMCPSMNL FSH Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 72%.
This gene encodes a testis-specific, differentiation antigen, acrosomal vesicle protein 1, that arises within the acrosomal vesicle during spermatogenesis, and is associated with the acrosomal membranes and matrix of mature sperm. This gene consists of 4 exons and its alternative splicing generates multiple distinct transcripts, which encode protein isoforms ranging from 81 to 265 amino acids. The longest transcript is the most abundant, comprising 53-72% of the total acrosomal vesicle protein 1 messages; the second largest transcript comprises 15-32%; the third and the fourth largest transcripts account for 3.4-8.3% and 8.7-12.5%, respectively; and the remaining transcripts combined account for < 1% of the total acrosomal vesicle protein 1 message. It is suggested that phenomena of cryptic splicing and exon skipping occur within this gene. The acrosomal vesicle protein 1 may be involved in sperm-zona binding or penetration, and it is a potential contraceptive vaccine immunogen for humans.
Specifications
| Acrosomal Vesicle Protein | |
| Polyclonal | |
| Unconjugated | |
| ACRV1 | |
| acrosomal protein SP-10; Acrosomal vesicle protein 1; Acrv1; D11S4365; Msa63; MSA-63; rSP10; Sp10; SP-10; SPACA2; sperm membrane protein; sperm protein 10 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 56 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P26436 | |
| ACRV1 | |
| Recombinant protein corresponding to Human Acrosomal Vesicle Protein. Recombinant protein control fragment (Product #RP-97353). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction