Learn More
Azlon™ Double Angled Beta Radiation Shield
Angled to allow comfortable working position when examining or handling samples
Brand:  Azlon SRS002
Product Code. 12078583
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
  • Shields in 10mm thick acrylic protect against beta radiation
  • Angled to allow comfortable working position when examining or handling samples
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Acrylic
Dual Angled Shield
445 mm
1
150 mm
10 mm