Learn More
Socorex™ Acura™ Manual 855 Multi-channel Micropipettes
Click to view available options
Volume (Metric):
0.5 to 10 μL
20 to 200 μL
40 to 350 μL
5 to 50 μL
Number of Channels:
8
12
Unit Size:
Each
Description
- Exceptional, user-friendly ergonomics.
- Smart and reliable volume adjustment.
- Precision digital display visible at all times.
- Justip™ adjustable tip ejector fitting most tips.
- Swift-set user calibration system.
- Shock, UV-light and autoclaving resistance.
- 360° rotation of volumetric module allows selection of optimal working position.
- Adjustable tip ejector.
Specifications
Specifications
| Accuracy | <±1.0% - <±0.8% |
| Certifications/Compliance | ISO 8655, GMP, GLP, NCCLS, CE directive IVD 98/79 EEC (in vitro diagnostic medical devices). |
| Autoclavable | Autoclavable |
| For Use With | Microplate |
| Model | Manual 855 |
| Number of Channels | 8 |
| Product Line | Acura |
| Type | Multi-Channel Micropipette |
| Volume (Metric) | 40 to 350 μL |
| Precision | <±0.6% - <±0.3% |
| Show More |