missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADAMTSL-1/Punctin Mouse anti-Human, Clone: 212125, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB1717-SP
This item is not returnable.
View return policy
Description
ADAMTSL-1/Punctin Monoclonal specifically detects ADAMTSL-1/Punctin in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| ADAMTSL-1/Punctin | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Q8N6G6 | |
| ADAMTSL1 | |
| Mouse myeloma cell line NS0-derived recombinant human ADAMTSL-1/Punctin Glu29-Ser525 Accession # Q8N6G6 | |
| 25 μg | |
| Primary | |
| Detects human ADAMTSL-1/Punctin in direct ELISAs and Western blots. | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| 212125 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ADAM-TS related protein 1, ADAMTSL1, ADAMTSL-1, ADAMTS-like 1, ADAMTS-like protein 1, ADAMTSR1MGC118805, C9orf94, chromosome 9 open reading frame 94, DKFZp686L03130, FLJ35283, FLJ41032, FLJ46891, MGC118803, MGC40193, PUNCTIN, Punctin-1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 92949 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction