Learn More
Eppendorf™ Adapter
Product Code. 15758379 Shop All Eppendorf Products
Click to view available options
For Use With (Equipment):
Adapter 13x75-100mm round bottom/PL/TB
Adapter 15mL conical tubes, PL/TB
Adapter 16x75-100mm round bottom/PL/TB
Adapter 17.5x100mm round bottom/PL/TB
Adapter 50mL conical tubes, PL/TB
for 500mL Nalgene
for 600mL TPP
Unit Size:
Pack of 2
Product Code. 15758379
Brand: Eppendorf™ 5920706004
Adaptors available for a variety of Eppendorf products to suit your needs.
Adapter, for 2.6 to 8mL , set of 2 pcs.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Adapter 13x75-100mm round bottom/PL/TB
No. per Pack Set of 2