Learn More
Thermo Scientific™ Adapters for TX-1000 Swinging Bucket Rotor
Product Code. 15149485 Shop All Thermo Scientific Products
Click to view available options
Capacity:
100 x 9/10 mL
150 to 300 mL Small Blood Bags
164 x 4.5/6 mL
20 x 50 mL
4 x 1000 mL
4 x 250 mL
4 x 500 mL
4 x 6 mL Standard or 4 x 2 mL Deep-Well
4 x 750 mL
48 x 50 mL
8 x 250 mL
96 x 15 mL
For Use With (Equipment):
1000mL Bio-Bottle
15mL Conical Tube
250mL Conical Bottle or 200mL Thermo Scientific Nunc Bottle or 175mL Thermo Scientific Nalgene Conical Bottle
250mL Thermo Scientific Nalgene Bottle
4.5/6mL Blood Collection Tube
500mL Corning Bottle
500mL Nalgene Bottle
50mL Double Biocontainment Vessel for 50mL Conical Tube (can be combined with ClickSeal lids)
50mL Thermo Scientific Nalgene Oak Ridge Tube
750 mL Polypropylene Wide-Mouth Bio-Bottle and any TX-750 Round adapter
9/10mL Blood Collection Tube
Microplate carriers
Small bag/cell culture bags 4 x 2 bags (< 350mL)
Unit Size:
4 sets
Each
Pack of 2
Pack of 4
Product Code. 15149485
Brand: Thermo Scientific™ 75004253
An assortment of adapters for outstanding flexibility
TRUSTED_SUSTAINABILITY
Specifications
Type Rotor Adapter
Tube Type 500 mL Nalgene Bottle
Capacity 4 x 500 mL
Max. Tube Diameter 70 mm
Max. Tube Length 160 mm
For Use With (Equipment) 500mL Nalgene Bottle