Learn More
Memmert™ Addition of Units for UF1060plus
Product Code. 17598865 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
UF1060plus Oven
Unit Size:
Each
Product Code. 17598865
Brand: Memmert™ UF1060plusFDAQ2
Only null left
Add to basket
Add to basket
Integration of one additional unit (up to max. 31 units) into an already existent FDA-software licence (only for units with TwinDISPLAY).
  • Up to max. 31 units
  • Only for units with TwinDISPLAY
Specifications
For Use With (Equipment) UF1060plus Oven
Type Unit Addition