missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Adenine sulfate, 98%, synthetic

Product Code. p-50837 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 10636562

Brand: Thermo Scientific Chemicals undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 321-30-2 | C10H12N10O4S | 368.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 321-30-2
Molecular Formula C10H12N10O4S
Molecular Weight (g/mol) 368.33
MDL Number MFCD00213655
InChI Key LQXHSCOPYJCOMD-UHFFFAOYSA-N
Synonym adenine sulfate, adenine hemisulfate, 7h-purin-6-amine sulfate 2:1, adeninsulfat, adeninium sulfate, adenine sulfate 2:1, adeninsulfat german, 1h-purin-6-amine, sulfate 2:1, diadenine sulphate, unii-741gjf3k9m
PubChem CID 9449
IUPAC Name 7H-purin-6-amine;sulfuric acid
SMILES OS(O)(=O)=O.NC1=C2NC=NC2=NC=N1.NC1=C2NC=NC2=NC=N1

Specifications

Absorbance 0.74 to 0.78 (A250/A260), 0.34 to 0.38 (A280/A260)
CAS Min % 97.5
CAS Max % 100.0
Melting Point 285.0°C
Color White to Yellow
Loss on Drying 2% max. (1 g, 105°C, 5 hrs) (vacuum)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Quantity 25 g
Beilstein 26, V,16, 100
Solubility Information Solubility in water: soluble
Spectrophotometry 97.5% min. (lambda max. 262nm,pH 1) molar absorptivity 13100+/-2%,on dry basis
Transmittance 95% min. (c=0.5, H2O, 430nm) 1cm cell
Formula Weight 184.17
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Adenine sulfate, 98%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.