missing translation for 'onlineSavingsMsg'
Learn More

Adenosine-5'-Diphosphate, MP Biomedicals™

Product Code. 11490232
Click to view available options
Quantity:
500 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Consists of the pyrophosphate group, the pentose sugar ribose, and the nucleobase adenine. Adenosine-5'-Diphosphate, MP Biomedicals is a nucleoside diphosphate that induces human platelet aggregation and non-competitively blocks the stimulated human plate

ADP (or 5’-ADP) is an ester of pyrophosphoric acid with the nucleosideadenosine. It is the product of ATP dephosphorylation by ATPases and it is converted back to ATP by ATP synthases. ATP is an important energy transfer molecule in cells.

ADP was used as a test compound for studying the endothelium-dependent vascular response in salt sensitive (DS) and salt resistant Dahl rats (DR). The product was used to study the different P2-purinergic receptor subtypes on canine vascular smooth muscle and endothelium.

TRUSTED_SUSTAINABILITY

Specifications

CAS 58-64-0
Molecular Formula C10H15N5O10P2
Quantity 500 mg
Synonym adenosine 5'-diphosphate, adenosine diphosphate, adp, adenosine-5'-diphosphate, adenosine pyrophosphate, adenosine 5'-trihydrogen diphosphate, adenosine-diphosphate, adenosine 5'-pyrophosphate, adp nucleotide, 5'-adenylphosphoric acid
Solubility Information Soluble in water (50mg/mL).
InChI Key XTWYTFMLZFPYCI-KQYNXXCUSA-N
SMILES C1=NC2=C(C(=N1)N)N=CN2C3C(C(C(O3)COP(=O)(O)OP(=O)(O)O)O)O
IUPAC Name [(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphono hydrogen phosphate
Molecular Weight (g/mol) 427.203
PubChem CID 6022
ChEBI CHEBI:16761
Formula Weight 427.2
Percent Purity ≥96%
Physical Form Powder
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'