missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adenosine-5'-diphosphate trilithium salt, 96 to 99%, For HPLC analysis, MP Biomedicals™
Click to view available options
Quantity:
1 g
5 g
Specifications
Specifications
Color | White |
Assay Percent Range | 96 to 99% |
Quantity | 1 g |
Formula Weight | 454 |
Physical Form | Crystalline Powder |
Chemical Name or Material | Adenosine-5'-diphosphate trilithium salt, For HPLC analysis |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'