Learn More
Color Coded High Resolution pH Buffer, Reagecon™
Shop All Reagecon Products
Click to view available options
Quantity:
500 mL
pH:
4
4.00
7
7.00
10.00
Unit Size:
500mL
Each
Description
- Colored pH Buffer Solutions
- High Resolution
- Tested at 20°C or 25°C
- Certified by Reagecon’s ISO/IEC 17025 AccreditedTest Method
- NIST traceable
- Presented in various pack sizes
Specifications
Specifications
| Melting Point | 0°C |
| Color | Yellow |
| pH | 7 |
| Physical Form | Liquid |
| Quantity | 500 mL |