Learn More
Color Coded High Resolution pH Buffer, Reagecon™
BIOPHARMA
Product Code. 15214643 Shop All Reagecon Products
Click to view available options
Quantity:
500 mL
pH:
4
4.00
7
7.00
10.00
Unit Size:
500mL
Each
Product Code. 15214643
Brand: Reagecon H70525C
Use color coded pH buffers for ease of use. Reagecon™ Color Coded High Resolution pH Buffers are tested at either 20°C or 25°C and are certified and NIST traceable.
  • Colored pH Buffer Solutions
  • High Resolution
  • Tested at 20°C or 25°C 
  • Certified by Reagecon’s ISO/IEC 17025 AccreditedTest Method
  • NIST traceable
  • Presented in various pack sizes
Specifications
Melting Point 0°C
Color Yellow
pH 7
Physical Form Liquid
Quantity 500 mL