Learn More
Brady™ Adhesive Polyester Safety Signs
Product Code. 11848030
Click to view available options
Width (Metric):
25 mm
50 mm
100 mm
200 mm
315 mm
Displayed Text:
Corrosive
DANGER of Suffocation
Oxidizer
Unit Size:
1 roll
Each
Product Code. 11848030
Brand: Brady™ 807788
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Certifications/Compliance RoHS compliant using EU Directive 2002/95/EC
Material Polyester
Quantity 1 Roll
Adhesive Type Permanent
Description Adhesive Polyester
For Use With (Application) Pipe markers and safety signs
Height (Metric) 25 mm
Width (Metric) 25 mm
Displayed Text Oxidizer
Product Type Safety Sign