missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ADHFE1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA531416
This item is not returnable.
View return policy
Description
Recommended positive controls: NT2D1, IMR32, U87-MG, MCF-7. Predicted reactivity: Mouse (89%), Rat (90%), Xenopus laevis (84%), Bovine (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The ADHFE1 gene encodes hydroxyacid-oxoacid transhydrogenase (EC 1.1.99.24), which is responsible for the oxidation of 4-hydroxybutyrate in mammalian tissues (Kardon et al., 2006 ).
Specifications
| ADHFE1 | |
| Polyclonal | |
| Unconjugated | |
| Adhfe1 | |
| 6330565B14Rik; ADH8; ADHFe1; AI043035; alcohol dehydrogenase 8; alcohol dehydrogenase iron-containing protein 1; alcohol dehydrogenase, iron containing 1; alcohol dehydrogenase, iron containing, 1; Fe-containing alcohol dehydrogenase; Fe-containing alcohol dehydrogenase 1; HMFT2263; HOT; hydroxyacid-oxoacid transhydrogenase, mitochondrial | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 137872 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q8IWW8 | |
| Adhfe1 | |
| Recombinant protein encompassing a sequence within the center region of human ADHFE1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto