missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adipic Acid, Free Acid, MP Biomedicals™
Click to view available options
Quantity:
500 g
Specifications
Specifications
CAS | 124-04-9 |
Molecular Formula | C6H10O4 |
Synonym | adipic acid, adipinic acid, 1,4-butanedicarboxylic acid, adilactetten, acifloctin, acinetten, 1,6-hexanedioic acid, molten adipic acid, kyselina adipova, adipinsaure german |
InChI Key | WNLRTRBMVRJNCN-UHFFFAOYSA-N |
SMILES | C(CCC(=O)O)CC(=O)O |
IUPAC Name | hexanedioic acid |
Molecular Weight (g/mol) | 146.142 |
PubChem CID | 196 |
ChEBI | CHEBI:30832 |
Formula Weight | 146.1 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'