missing translation for 'onlineSavingsMsg'
Learn More

Adoprazine, MedChemExpress

Product Code. 16438767
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16438767

Brand: Medchem Express HY14782

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Adoprazine (SLV313) is a full 5-HT1A receptor agonist with a pEC50 of 9 at cloned h5-HT1A receptors. Adoprazine (SLV313) is a full D2 and D3 receptor antagonist with pA2s of 9.3 and 8.9 at hD2 and hD3 receptors, respectively. Adoprazine (SLV313) has the characteristics of atypical antipsychotics.

Adoprazine (SLV313) is a full 5-HT1A receptor agonist with a pEC50 of 9 at cloned h5-HT1A receptors. Adoprazine (SLV313) is a full D2 and D3 receptor antagonist with pA2s of 9.3 and 8.9 at hD2 and hD3 receptors, respectively. Adoprazine (SLV313) has the characteristics of atypical antipsychotics.

Specifications

Chemical Name or Material Adoprazine
CAS 222551-17-9
Color Yellow
Purity Grade Notes Research
Molecular Formula C24H24FN3O2
Quantity 5 mg
Synonym SLV313
Solubility Information DMSO : 50 mg/mL (123.32 mM; Need ultrasonic)
SMILES FC1=CC=C(C2=CN=CC(CN3CCN(C4=C5OCCOC5=CC=C4)CC3)=C2)C=C1
Molecular Weight (g/mol) 405.46
Formula Weight 405.46
Percent Purity 98.1%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.