Learn More
Advanced Instruments Osmolality Linearity Set
For monitoring performance of any osmometer in the range 0-2000 mOsm/kg
Brand:  Advanced Instruments 3LA028
Encompass_Preferred
Product Code. 15705682
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Each level of standard supplied in the set should be run and documented to verify the reportable range of patient results
  • Run the linearity set until accurate results obtained or recalibration needed
  • For in vitro diagnostic use with all brands of osmometers
Specifications
Specifications
CLIA, CE
100, 500, 900, 1500, and 2000 mOsm/kg H2O
Osmolality Linearity set
2 x 5 mL
2°C to 30°C
3320 Micro-osmometer parts and supplies
Monitoring performance of any osmometer in the range 0 to 2000mOsm/kg