Learn More
Melford Agarose Low EEO
Molecular Biology Grade.
Brand:  Melford A20080-1000.0
 View more versions of this product
Product Code. 30017976
  • 1905.00 EUR / 1kg
Description
Molecular Biology Grade.
Molecular Biology Grade. High purity for preparative and analytical electrophoresis of nucleic acids >1000 bp.
Gelling Temperature:
34.5 -37.5°C
Sulfate:
0.15
EEO:
0.09- 0.13
DNA Binding:
None Detected
DNA and RNase:
None Detected
Specifications
Specifications
Agarose, Low EEO
9012-36-6
1 kg
Molecular Biology