accessibility menu, dialog, popup
Test
Search
Order Status
Quick Order
Support
Sign In
chevron-down
chevron-down
Learn More
Home
Products
Mixers
Mortars and Pestles
Giese™ Agate Pestle
11766794
Print
Giese™ Agate Pestle
Length: 50mm
Brand:
Giese™ PESTLE50
View more versions of this product
Product Code.
11766794
26.00 EUR
/ Each
Quantity
Estimated Shipment: 16-01-2025
Log in
to see stock.
Explore more special offers
Add to basket
This item is not returnable.
View return policy
Specifications
Length (Metric)
50 mm