Learn More
Brady™ Aggressive Adhesive Multipurpose Nylon Labels
Product Code. 17940680
Click to view available options
Dimensions (LxW):
12.70 mm (W) x 4.88 m (L)
19.05 mm (W) x 12.70 mm (H)
19.05 mm (W) x 4.88 m (L)
20.96 mm (W) x 9.53 mm (H)
22.86 mm (W) x 12.70 mm (H)
25.40 mm (W) x 12.70 mm (H)
25.40 mm (W) x 4.88 m (L)
38.10 mm (W) x 31.75 mm (H)
38.10 mm (W) x 4.88 m (L)
48.26 mm (W) x 25.40 mm (H)
9.53 mm (W) x 4.88 m (L)
Format:
Continuous
Die-cut
Unit Size:
Each
Pack of 100
Pack of 160
Pack of 180
Pack of 200
Pack of 220
Product Code. 17940680
Brand: Brady™ 170793
Only null left
Add to basket
Add to basket
The B-499 nylon cloth label is made of tough and versatile, general-purpose label material. Print almost anywhere with a Brady™ portable label printer.
  • Ideal for labeling tightly-curved surfaces such as wires and cables, as well as general-purpose lab, equipment panels, faceplates and terminal blocks
  • Designed to withstand extreme temperatures, chemicals, greases, oils, cleaners and detergents
  • Permanent adhesive labels that stick to smooth, rough and powder-coated surfaces alike
Specifications
Certifications/Compliance UL Recognised;CSA Accepted;Halogen Free (DIN VDE 0472/part 815) Approvals are for White material only Compliant with Directive 2011/65/EU and 2015/863
Adhesive Type Permanent Acrylic
Color White
Finish Matte
Format Continuous
Material Nylon Cloth
Product Type Label
Sterility Non-sterile
For Use With (Application) Component and equipment labelling;Data and telecommunications labelling;Electrical labelling;General identification;Wire and cable labelling
For Use With (Equipment) BMP41;BMP51;BMP53;M511;M410;M510
Quantity 1
Temperature Range (English) -40°F to 194°F
Temperature Range (Metric) -40°C to 90°C
Thickness (English) 0.0065 in
Thickness (Metric) 0.166 mm
Dimensions (LxW) 9.53 mm (W) x 4.88 m (L)
Length (English) 16 ft.
Length (Metric) 4.88 m
Width (English) 0.375 in
Width (Metric) 9.53 mm
Alternate Part Number M4C-375-499