missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AGPAT6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA526651
This item is not returnable.
View return policy
Description
This antibody is predicted to react with bovine based on sequence homology.
Lysophosphatidic acid acyltransferases (EC 2.3.1.51) catalyze the conversion of lysophosphatidic acid (LPA) to phosphatidic acid (PA). LPA and PA are involved in signal transduction and lipid biosynthesis.
Specifications
| AGPAT6 | |
| Polyclonal | |
| Unconjugated | |
| GPAT4 | |
| 1-acylglycerol-3-phosphate O-acyltransferase 6; 1-acylglycerol-3-phosphate O-acyltransferase 6 (lysophosphatidic acid acyltransferase, zeta); 1-acyl-sn-glycerol-3-phosphate acyltransferase zeta; 1-AGP; 1-AGP acyltransferase 6; 1-AGPAT 6; acyl-CoA:glycerol-3-phosphate acyltransferase 4; AGPAT6; AU041707; AW545732; Glycerol-3-phosphate acyltransferase 4; glycerol-3-phosphate acyltransferase 6; glycerol-3-phosphate acyltransferase-4; Gpat4; LPAATZ; LPAAT-zeta; lysophosphatidic acid acyltransferase zeta; putative lysophosphatidic acid acyltransferase; RGD1310520; testis spermatogenesis apoptosis-related protein 7; TSARG7; UNQ551/PRO1108 | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 102247, 137964 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q86UL3, Q8K2C8 | |
| GPAT4 | |
| KLH conjugated synthetic peptide between 45-74 amino acids from the N-terminal region of human AGPAT6. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction