Learn More
Tocris Bioscience™ AH 7614
Selective FFA4 (GPR120) antagonist
Brand:  Tocris Bioscience™ 5256/10
 View more versions of this product
Product Code. 18767976
  • 167.00 EUR / 10mg
[resources not loaded]
Explore more special offers
Description
Description
AH 7614 is a selective free fatty acid receptor 4 (FFA4/GPR120) antagonist (pIC50 values are 7.1 and <4.6 for human FFA4 and FFA1 receptors respectively). Inhibits linoleic acid and GSK 137647A -induced intracellular calcium accumulation in U2OS osteosarcoma cells expressing the FFA4 receptor.
Specifications
Specifications
>98%
AH 7614
AH 7614
10 mg
351.42