Learn More
Tocris Bioscience™ AH 7614
Selective FFA4 (GPR120) antagonist
Brand: Tocris Bioscience™ 5256/10
This item is not returnable.
View return policy
Description
AH 7614 is a selective free fatty acid receptor 4 (FFA4/GPR120) antagonist (pIC50 values are 7.1 and <4.6 for human FFA4 and FFA1 receptors respectively). Inhibits linoleic acid and GSK 137647A -induced intracellular calcium accumulation in U2OS osteosarcoma cells expressing the FFA4 receptor.Specifications
>98% | |
AH 7614 | |
AH 7614 |
10 mg | |
351.42 |
Product Content Correction
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction