Learn More
Lenz Laborglasintrumente Air Leak Tubewith Bell
Click to view available options
Length (Metric):
180 mm
Unit Size:
Each
Description
- Air leak tube with bell
- To avoid boiling delay
- Dimensions: L 180 x 4 mm dia.
- Made of DURAN™ tubing
Specifications
Specifications
| Length (Metric) | 180 mm |
| Diameter (Metric) Outside | 4 mm |