Learn More
Syngene™ Airclean 600 Dead Air Box
GREENER_CHOICE
Product Code. 16554341 Shop All Syngene Products
Click to view available options
Width (English):
32 in.
48 in.
Unit Size:
Each
Product Code. 16554341
Brand: Syngene™ SYN48DBCE
Airclean 600 dead air box equipped with a UVTect Controller.
  • With UVTect Controller
  • Price includes fluorescent lights, UV lights, UV shelf with pipette holder, and slip hatch
Specifications
Product Type Dead Air Box
Certifications/Compliance AC UL, CSA, and CE approved
Includes UV Controller
Voltage 220 V
Width (English) 48 in.
Width (Metric) 121.92 cm