Learn More
Al-23 Grinding Pieces, H: 10mm, 440 Pieces/Kg, Thermo Scientific Chemicals
1073.00 EUR
Specifications
Chemical Name or Material Al-23 Grinding Pieces
Form Grinding Pieces
View More Specs
Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15431307
View Documents Promotion Details
Thermo Scientific Alfa Aesar
033190.A1
1 kg
1073.00 EUR
1kg
View alternative products
Save Up to 
»
Description
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Al-23 Grinding Pieces
Grinding Pieces
Videos
Safety and Handling
Safety and Handling
missing translation for 'tsca' : U
Recommended Storage : Ambient temperatures