Learn More
Alfuzosin, MedChemExpress
MedChemExpress Alfuzosin is an α1 adrenergic receptor antagonist used to treat benign prostatic hyperplasia (BPH).
Brand:  Medchem Express HY-B0192
Encompass_Preferred
 View more versions of this product
Product Code. 16474287
  • 209.25 EUR / 50mg
Description
Alfuzosin is an α1 adrenergic receptor antagonist used to treat benign prostatic hyperplasia (BPH).
Specifications
Specifications
Alfuzosin
White
C19H27N5O4
SL 77499
O=C(C1OCCC1)NCCCN(C2=NC(N)=C3C=C(OC)C(OC)=CC3=N2)C
389.45
Research
81403-80-7
Research
50 mg
DMSO : ≥ 50 mg/mL (128.39 mM)
389.45
99.67%
Solid