Learn More
Alfuzosin, MedChemExpress
MedChemExpress Alfuzosin is an α1 adrenergic receptor antagonist used to treat benign prostatic hyperplasia (BPH).
Brand: Medchem Express HY-B0192
This item is not returnable.
View return policy
Description
Alfuzosin is an α1 adrenergic receptor antagonist used to treat benign prostatic hyperplasia (BPH).Specifications
Alfuzosin | |
White | |
C19H27N5O4 | |
SL 77499 | |
O=C(C1OCCC1)NCCCN(C2=NC(N)=C3C=C(OC)C(OC)=CC3=N2)C | |
389.45 | |
Research |
81403-80-7 | |
Research | |
50 mg | |
DMSO : ≥ 50 mg/mL (128.39 mM) | |
389.45 | |
99.67% | |
Solid |