missing translation for 'onlineSavingsMsg'
Learn More

Alkaline Phosphatase, MP Biomedicals™

366.00 EUR - 1470.00 EUR

Specifications

CAS 9001-78-9
Content And Storage Store at 0°C.
Format Solid
Molecular Weight (g/mol) 496.569
Product Type Alkaline Phosphatase
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11481431
View Documents
MP Biomedicals
0215934705
5mg
366.00 EUR
5mg
Please to purchase this item. Need a web account? Register with us today!
11491431
View Documents
MP Biomedicals
0215934710
10mg
675.00 EUR
10mg
Please to purchase this item. Need a web account? Register with us today!
11401441
View Documents
MP Biomedicals
0215934725
25mg
1470.00 EUR
25mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

9001-78-9
Solid
Alkaline Phosphatase
Store at 0°C.
496.569
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.