missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha 2-Macroglobulin Mouse anti-Human, Clone: A2M/3623, Novus Biologicals™
Mouse Monoclonal Antibody
240.81 EUR
Specifications
Antigen | alpha 2-Macroglobulin |
---|---|
Clone | A2M/3623 |
Concentration | 0.2 mg/ml |
Dilution | Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array |
Applications | Immunohistochemistry (Paraffin), Peptide Array |
Description
alpha 2-Macroglobulin Monoclonal specifically detects alpha 2-Macroglobulin in Human samples. It is validated for Immunohistochemistry-Paraffin, Protein Array.Specifications
alpha 2-Macroglobulin | |
0.2 mg/ml | |
Immunohistochemistry (Paraffin), Peptide Array | |
Unconjugated | |
Mouse | |
Neuroscience | |
10 mM PBS with 0.05% BSA | |
2 | |
IgG1 κ | |
Protein A or G purified |
A2M/3623 | |
Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
Monoclonal | |
Purified | |
RUO | |
Human | |
alpha-2-M, alpha-2-macroglobulin, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 5, CPAMD5DKFZp779B086, FWP007, S863-7 | |
Human recombinant alpha 2-Macroglobulin protein fragment (around aa 604-748) (exact sequence is proprietary) | |
Primary | |
Store at 4°C. |