Learn More
alpha-Bisabolol, 96%, Thermo Scientific Chemicals
61.30 EUR - 193.00 EUR
Chemical Identifiers
CAS | 515-69-5 |
---|---|
Molecular Formula | C15H26O |
Molecular Weight (g/mol) | 222.372 |
MDL Number | MFCD03846910 |
InChI Key | RGZSQWQPBWRIAQ-LSDHHAIUSA-N |
Synonym | bisabolol, alpha-bisabolol, +-alpha-bisabolol, d-alpha-bisabolol, .alpha.-bisabolol, +-1'r,2r-alpha-bisabolol, 2r-6-methyl-2-1r-4-methylcyclohex-3-en-1-yl hept-5-en-2-ol, 3-cyclohexene-1-methanol, .alpha.,4-dimethyl-.alpha.-4-methyl-3-pentenyl-, r*,r*, dsstox_cid_25964, dsstox_rid_81260 |
PubChem CID | 1549992 |
IUPAC Name | (2R)-6-methyl-2-[(1R)-4-methylcyclohex-3-en-1-yl]hept-5-en-2-ol |
SMILES | CC1=CCC(CC1)C(C)(CCC=C(C)C)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11406974
|
Thermo Scientific Alfa Aesar
B21457.09 |
10 g |
61.30 EUR
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11416974
|
Thermo Scientific Alfa Aesar
B21457.18 |
50 g |
193.00 EUR
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It finds its application as a masking agent, skin conditioning agent and soothing agent in cosmetics industry. The most important effects of Bisabolol for the use in cosmetics are anti-inflammatory, wound-healing, anti-bacterial and anti-mycotic. Bisabolol is therefore perfectly suited for the use in all kinds of skin-care products It can be used as active substance in cosmetic preparations for the protection and care of sensitive skin, preparations for babies and children, sunscreen and after-sun products, aftershaves and preparations for the oral hygiene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt finds its application as a masking agent, skin conditioning agent and soothing agent in cosmetics industry. The most important effects of Bisabolol for the use in cosmetics are anti-inflammatory, wound-healing, anti-bacterial and anti-mycotic. Bisabolol is therefore perfectly suited for the use in all kinds of skin-care products It can be used as active substance in cosmetic preparations for the protection and care of sensitive skin, preparations for babies and children, sunscreen and after-sun products, aftershaves and preparations for the oral hygiene.
Solubility
Soluble in alcohol, natural, mineral and synthetic oils. Insoluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
515-69-5 | |
222.372 | |
RGZSQWQPBWRIAQ-LSDHHAIUSA-N | |
1549992 | |
CC1=CCC(CC1)C(C)(CCC=C(C)C)O |
C15H26O | |
MFCD03846910 | |
bisabolol, alpha-bisabolol, +-alpha-bisabolol, d-alpha-bisabolol, .alpha.-bisabolol, +-1'r,2r-alpha-bisabolol, 2r-6-methyl-2-1r-4-methylcyclohex-3-en-1-yl hept-5-en-2-ol, 3-cyclohexene-1-methanol, .alpha.,4-dimethyl-.alpha.-4-methyl-3-pentenyl-, r*,r*, dsstox_cid_25964, dsstox_rid_81260 | |
(2R)-6-methyl-2-[(1R)-4-methylcyclohex-3-en-1-yl]hept-5-en-2-ol |
Specifications
515-69-5 | |
135°C (275°F) | |
1.495 | |
5733954 | |
14,1241 | |
Soluble in alcohol,natural,mineral and synthetic oils. Insoluble in water. | |
CC1=CCC(CC1)C(C)(CCC=C(C)C)O | |
222.372 | |
222.37 | |
Liquid |
0.93 | |
C15H26O | |
MFCD03846910 | |
Air Sensitive | |
bisabolol, alpha-bisabolol, +-alpha-bisabolol, d-alpha-bisabolol, .alpha.-bisabolol, +-1'r,2r-alpha-bisabolol, 2r-6-methyl-2-1r-4-methylcyclohex-3-en-1-yl hept-5-en-2-ol, 3-cyclohexene-1-methanol, .alpha.,4-dimethyl-.alpha.-4-methyl-3-pentenyl-, r*,r*, dsstox_cid_25964, dsstox_rid_81260 | |
RGZSQWQPBWRIAQ-LSDHHAIUSA-N | |
(2R)-6-methyl-2-[(1R)-4-methylcyclohex-3-en-1-yl]hept-5-en-2-ol | |
1549992 | |
96% | |
Bisabolol |
Safety and Handling
missing translation for 'einecsNumber' : 208-205-9
missing translation for 'rtecsNumber' : MJ9685000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only