Learn More
KGW Isotherm™ Dish-Shaped Dewar Flasks
Product Code. p-8008294
Click to view available options
Capacity (Metric):
260 mL
400 mL
800 mL
5300 mL
Unit Size:
Each
Product Code. 10790972
Brand: KGW Isotherm™ 1195
Only null left
Add to basket
Add to basket
For the cooling of small samples. KGW Isotherm™ Dish-Shaped Dewar Flasks are suitable as a hot or cold bath.
  • Easy handling
  • Suitable on magnetic stirrer
  • Full silvered
  • Made of borosilicate glass 3.3
  • Polyethylene lid (not included)
Specifications
Cap Lid Type Lid not included
Color Silver
Depth (Metric) Inside 80mm
Diameter (Metric) Outer 170mm
For Use With (Application) Laboratory technique, Medicinal technique
Height (Metric) 125mm
Base Material Borosilicate Glass
Capacity (Metric) 800 mL
Material Aluminum