missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Aluminum Evacuation Panel Safety Signs
Brand: Brady™ 807235
This item is not returnable.
View return policy
Specifications
Aluminum | |
with Down Arrow | |
297 mm | |
148 mm | |
Evacuation Panel Safety Sign |
1 Ea. | |
for Indoor and Outdoor Safety Signs | |
120°C | |
Exit and emergency exit | |
White on Green |