Learn More
Wheaton™ Aluminum Fully Tear Off Seal
Brand: Wheaton™ 700009
55.10 EUR valid until 2024-12-31
Use promo code "23259" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
Silver | |
Aluminum | |
Closure | |
20 mm |