Learn More
Mettler Toledo™ Aluminum Standard Crucible
Brand:  Mettler Toledo™ 00026763
Product Code. 15456091
  • 692.00 EUR / Each
Estimated Shipment: 28-02-2025
to see stock.
Explore more special offers
Specifications
Aluminum
1Pack
6mm
Standard Crucible
640°C
40 μL
1.6mm