missing translation for 'onlineSavingsMsg'
Learn More

AM-92016 hydrochloride, MedChemExpress

Product Code. 16463308
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16463308

Brand: Medchem Express HY101253

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AM-92016 hydrochloride is a specific blocker of rectifier potassium current (IK). AM-92016 hydrochloride delays rectifier potassium channel (IK), repolarizes the membrane thereby restricting the duration of the nerve impulse thereby restricting the duration of the nerve impulse.

AM-92016 hydrochloride is a specific blocker of rectifier potassium current (IK). AM-92016 hydrochloride delays rectifier potassium channel (IK), repolarizes the membrane thereby restricting the duration of the nerve impulse thereby restricting the duration of the nerve impulse.

Specifications

Chemical Name or Material AM-92016 hydrochloride
CAS 133229-11-5
Color Yellow
Purity Grade Notes Research
Molecular Formula C19H25Cl3N2O4S
Quantity 10 mg
Solubility Information DMSO : 260 mg/mL (537.37 mM; Need ultrasonic)
SMILES CS(=O)(NC1=CC=C(OCC(O)CN(CCC2=CC=C(Cl)C(Cl)=C2)C)C=C1)=O.Cl
Molecular Weight (g/mol) 483.84
Formula Weight 483.84
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.