missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amberlyst 15, MP Biomedicals™
Soluble in water
95.85 EUR - 424.00 EUR
Identifiants chimiques
CAS | 9037-24-5 |
---|---|
Molecular Formula | MFCD00145841 |
Molecular Weight (g/mol) | 0.00 |
MDL Number | MFCD00145841 |
InChI Key | YZUPZGFPHUVJKC-UHFFFAOYSA-N |
Synonym | 2-bromoethyl methyl ether, 2-methoxyethyl bromide, ethane, 1-bromo-2-methoxy, ether, 2-bromoethyl methyl, methoxyethyl bromide, 2-bromoethylmethylether, bromoethyl methyl ether, 2-bromoethylmethyl ether, 2-bromoethylmethyl-ether, 2-bromo-1-methoxyethane |
PubChem CID | 80972 |
IUPAC Name | 1-bromo-2-methoxyethane |
SMILES | * |
Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
11433141
|
MP Biomedicals
0215033680 |
100 g |
95.85 EUR
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11443141
|
MP Biomedicals
0215033690 |
500 g |
424.00 EUR
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Identifiants chimiques
9037-24-5 | |
0.00 | |
YZUPZGFPHUVJKC-UHFFFAOYSA-N | |
80972 | |
* |
MFCD00145841 | |
MFCD00145841 | |
2-bromoethyl methyl ether, 2-methoxyethyl bromide, ethane, 1-bromo-2-methoxy, ether, 2-bromoethyl methyl, methoxyethyl bromide, 2-bromoethylmethylether, bromoethyl methyl ether, 2-bromoethylmethyl ether, 2-bromoethylmethyl-ether, 2-bromo-1-methoxyethane | |
1-bromo-2-methoxyethane |
Spécification
Small Beads | |
MFCD00145841 | |
Brown/Green | |
YZUPZGFPHUVJKC-UHFFFAOYSA-N | |
1-bromo-2-methoxyethane | |
80972 |
9037-24-5 | |
MFCD00145841 | |
2-bromoethyl methyl ether, 2-methoxyethyl bromide, ethane, 1-bromo-2-methoxy, ether, 2-bromoethyl methyl, methoxyethyl bromide, 2-bromoethylmethylether, bromoethyl methyl ether, 2-bromoethylmethyl ether, 2-bromoethylmethyl-ether, 2-bromo-1-methoxyethane | |
* | |
0.00 | |
Amberlyst 15(H) |