Learn More
Invitrogen™ RNase-free Tips, 1,000 μL size
Provided in ten (10) racks containing 100 tips each. Invitrogen™ Ambion™ RNase-free Tips, 1000μL size are subjected to rigorous testing for RNase contamination and are certified to be RNase- and DNase-free.
Brand: Invitrogen™ AM12660
Additional Details : CAS Number : 9048-46-8 Weight : 2.51000kg
Description
Invitrogen™ Tips are compatible with Eppendorf™, Finnpipette, Oxford Benchmate™, Pipetman™, Proline™, Rainin, Socorex, and other brands of pipettors.
Specifications
1000 μL | |
Sterile | |
Store at room temperature. | |
Universal Fit |
Non-Filtered | |
RNase-free Tips | |
Room Temperature | |
Ambion |