missing translation for 'onlineSavingsMsg'
Learn More

AMG-47a, MedChemExpress

Product Code. 16467498
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16467498

Brand: Medchem Express HY18303

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AMG-47a is a potent and orally active lymphocyte-specific protein tyrosine kinase (Lck) inhibitor, with an IC50 of 0.2 nM. AMG-47a also inhibits VEGF2, p38α, Jak3 and MLR and IL-2 with IC50s of 1 nM, 3 nM, 72 nM, 30 nM and 21 nM, respectively. AMG-47a has anti-inflammatory activity.

AMG-47a is a potent and orally active lymphocyte-specific protein tyrosine kinase (Lck) inhibitor, with an IC50 of 0.2 nM. AMG-47a also inhibits VEGF2, p38α, Jak3 and MLR and IL-2 with IC50s of 1 nM, 3 nM, 72 nM, 30 nM and 21 nM, respectively. AMG-47a has anti-inflammatory activity.

Specifications

Chemical Name or Material AMG-47a
Physical Form Solid
CAS 882663-88-9
Color Light Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C29H28F3N5O2
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : ≥ 34 mg/mL (63.48 mM)
SMILES O=C(NC1=CC=CC(C(F)(F)F)=C1)C2=CC=C(C)C(C3=CC4=CN=C(NCCN5CCOCC5)N=C4C=C3)=C2
Molecular Weight (g/mol) 535.56
Formula Weight 535.56
Percent Purity 98.72%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.