missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals Amino Acid Diet, Synthetic, Pellet
Diet rich in Amino Acids. Can be added into agar gel to increase palatability.
1655.00 EUR - 6040.00 EUR
Specifications
Product Type | Animal Diet |
---|---|
Autoclavable | Autoclavable |
Disposable | Single-use |
Format | Pellet |
Sterility | Non-sterile |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11467341
|
MP Biomedicals
0296036210 |
10 kg |
1655.00 EUR
10kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11477341
|
MP Biomedicals
0296036220 |
20 kg |
2845.00 EUR
20kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11487341
|
MP Biomedicals
0296036250 |
50 kg |
6040.00 EUR
50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Diet rich in amino acids
- Can be added into agar gel to increase palatability
- Can be used for research
Specifications
Animal Diet | |
Single-use | |
Non-sterile | |
1 ft. |
Autoclavable | |
Pellet | |
Refrigerated | |
0.3048 m |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MP Biomedicals Amino Acid Diet, Synthetic, Pellet