missing translation for 'onlineSavingsMsg'
Learn More

Ammonia, 25% w/w, Reagent Grade, Scharlab

Product Code. 11319823
Click to view available options
Quantity:
1 L
2.5 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Laboratory reagent for use in analytical chemistry.

  • Synonyms: Ammonia water, Ammonium hydroxide solution
  • Molecular Formula: NH3
  • Molar Mass: 17.03g/mol
  • CAS: 1336-21-6
  • EIN ECS No.: 215-647-6

Chemical Identifiers

CAS 1336-21-6
Molecular Formula H5NO
Molecular Weight (g/mol) 35.05
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
Synonym ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
PubChem CID 14923
ChEBI CHEBI:18219
IUPAC Name amine hydrate
SMILES N.O

Specifications

Boiling Point 37.7°C
Melting Point -57.5°C
Color Colorless
Physical Form Liquid
Quantity 2.5 L
Formula Weight 17.03g/mol
Percent Purity 25 to 30% (acidimetric, NH3)
Grade Reagent
Vapor Pressure ∼500hPa at 20°C
Density 0.90g/cm3
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ammonia, solution 25% w/w, reagent grade, Reag. Ph Eur
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • ACUTE AQUATIC Acute 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contains : Ammonia
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'