Learn More
WTW™ Ammonia and Ammonium Test Kits
Click to view available options
Type:
Ammonium (NH4) Reagent Test Kit
Calcium Reagent Test Kit
Cuvette
Sulfate (SO4) Reagent Test Kit
Unit Size:
Each
Pack of 100
Pack of 25
Pack of 50
Specifications
Specifications
| For Use With (Application) | For waste water, drinking water, sea water |
| For Use With (Equipment) | For PhotoLab™ S12, PhotoLab™ Spectral photometers |
| Includes | Certificates |
| Quantity | 25 Pack |
| Range | 2 to 150 mg/L NH4-N, 2.6 to 193 mg/L NH4+ |
| Test Method | Photometric |
| Type | Cuvette |