missing translation for 'onlineSavingsMsg'
Learn More

Ammonium chloride, BAKER, J.T. Baker™

Product Code. 10497460
Click to view available options
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Meets Reagent Specifications for testing Ph. Eur. Monographs

Specifications

Melting Point 335°C
CAS 12125-02-9
pH 4.6 to 6.0 (5% Solution at 25°C)
Quantity 1 kg
Assay Percent Range 99.5 to 100.5%
Molecular Formula ClH4N
MDL Number MFCD00011420
InChI Key NLXLAEXVIDQMFP-UHFFFAOYSA-N
SMILES N.Cl
IUPAC Name amine hydrochloride
Molecular Weight (g/mol) 53.49
Identification Passes Test
Loss on Drying max. 0.5%
Packaging Plastic bottle for solids
Density 1.5256 g/cm³ (20°C)
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'