missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Hydroxide Solution, 28 to 30%, ACS Reagent, Honeywell Fluka™
ACS Reagent, 28.0-30.0% NH3 basis
44.28 EUR - 171.76 EUR
Chemical Identifiers
CAS | 1336-21-6 |
---|---|
Molecular Formula | H5NO |
Molecular Weight (g/mol) | 35.05 |
MDL Number | MFCD00066650 |
InChI Key | VHUUQVKOLVNVRT-UHFFFAOYSA-N |
Synonym | ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh |
PubChem CID | 14923 |
ChEBI | CHEBI:18219 |
SMILES | N.O |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
1336-21-6 | |
35.05 | |
VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
14923 | |
N.O |
H5NO | |
MFCD00066650 | |
ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh | |
CHEBI:18219 |
Specifications
1336-21-6 | |
NH4OH | |
MFCD00066650 | |
3587154 | |
ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh | |
N.O | |
0.27 | |
14923 | |
35.05g/mol | |
≤0.002% | |
115mmHg (20°C) |
H5NO | |
28 to 30% NH3 basis | |
2672 | |
0.9g/mL at 25°C | |
VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
ammonium hydroxide | |
35.05 | |
CHEBI:18219 | |
ACS Reagent | |
1.2 (vs air) | |
Ammonium Hydroxide Solution |
Safety and Handling
P273-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338-P391
H302-H314-H335-H400