missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Persulfate, Reagent grade, 98%, Honeywell™
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Poly Bottle
Description
- ≥95% for general laboratory use.
Specifications
Specifications
Melting Point | 120°C |
Color | White |
Quantity | 2.5 kg |
Linear Formula | (NH4)2S2O8 |
UN Number | UN1444 |
Formula Weight | 228.2g/mol |
Percent Purity | 98% |
Grade | Reagent |
Packaging | Poly Bottle |
Appearance | Powder/Crystals/Chunks |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction