missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Persulfate, Reagent grade, 98%, Honeywell™

Product Code. 15672000
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Reagent Grade, 98%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7727-54-0
Molecular Formula H8N2O8S2
Molecular Weight (g/mol) 228.19
MDL Number MFCD00003390
InChI Key ROOXNKNUYICQNP-UHFFFAOYSA-N
Synonym ammonium persulfate, ammonium peroxydisulfate, diammonium peroxydisulfate, diammonium persulfate, ammoniumpersulfate, diammonium peroxodisulphate, diammonium peroxydisulphate, unii-22qf6l357f, ccris 1430, persulfate d'ammonium french
PubChem CID 62648
IUPAC Name diazanium;sulfonatooxy sulfate
SMILES [NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O

Specifications

Melting Point 120°C
Color White
Quantity 500 g
Linear Formula (NH4)2S2O8
UN Number UN1444
Formula Weight 228.2g/mol
Percent Purity 98%
Grade Reagent
Packaging Poly Bottle
Appearance Powder/Crystals/Chunks
Density 1.98g/cu.cm; Vapor Density: 7.9 (vs air)
Chemical Name or Material Ammonium Persulfate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.