missing translation for 'onlineSavingsMsg'
Learn More

Ammonium sulfate, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99%, Honeywell Fluka™

Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥99%

57.97 EUR - 1194.36 EUR

Chemical Identifiers

CAS 7783-20-2
Molecular Formula H8N2O4S
Molecular Weight (g/mol) 132.13
MDL Number MFCD00003391
InChI Key BFNBIHQBYMNNAN-UHFFFAOYSA-N
Synonym ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate
PubChem CID 6097028
ChEBI CHEBI:62946
IUPAC Name diazanium;sulfate
SMILES N.N.OS(O)(=O)=O
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15623240
View Documents
Honeywell-Fluka
31119-1KG
1 kg
57.97 EUR
1kg
Please to purchase this item. Need a web account? Register with us today!
15653240
View Documents
Honeywell-Fluka
31119-5KG
5 kg
241.54 EUR
5kg
Please to purchase this item. Need a web account? Register with us today!
15633240
View Documents
Honeywell-Fluka
31119-4X5KG
4 x 5 kg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15643240
View Documents
Honeywell-Fluka
31119-50KG
50 kg
1194.36 EUR
50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

7783-20-2
132.13
BFNBIHQBYMNNAN-UHFFFAOYSA-N
6097028
diazanium;sulfate
H8N2O4S
MFCD00003391
ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate
CHEBI:62946
N.N.OS(O)(=O)=O

Specifications

7783-20-2
(NH4)2SO4
NONH for all modes of transport
BFNBIHQBYMNNAN-UHFFFAOYSA-N
diazanium;sulfate
6097028
132.14g/mol
ACS/puriss. p.a.
1.77g/mL at 25°C
H8N2O4S
MFCD00003391
ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate
N.N.OS(O)(=O)=O
132.13
CHEBI:62946
≥99%
≤0.005% (as SO4)
Ammonium Sulfate, Reag. Ph. Eur. reag. ISO
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 231-984-1

missing translation for 'rtecsNumber' : BS4500000

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.